Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113131.5 | NP_188872.2 | 2 - 3 | 654 | 92 | |
GenBank mRNA | AY062502.1 | - | 2 - 3 | 546 | 92 | |
AY093256.1 | - | 2 - 3 | 521 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |