Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113189.5 | NP_188929.1 | 4 - 5 | 1484 | 76 | |
GenBank mRNA | AF370492.1 | - | 4 - 5 | 1375 | 76 | |
AK226559.1 | - | 4 - 5 | 1333 | 76 | ||
AY050869.1 | - | 4 - 5 | 1321 | 76 | ||
AY054169.1 | - | 4 - 5 | 1333 | 76 | ||
AY064648.1 | - | 4 - 5 | 1261 | 76 | ||
AY091207.1 | - | 4 - 5 | 1261 | 76 | ||
BT002538.1 | - | 4 - 5 | 1332 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |