Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338655.1 | NP_001327897.1 | 4 - 5 | 824 | 131 | |
NM_113284.3 | NP_189022.1 | 5 - 6 | 1148 | 131 | ||
GenBank mRNA | AK117618.1 | - | 5 - 6 | 940 | 131 | |
BT005102.1 | - | 5 - 6 | 893 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |