Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113363.2 | NP_189095.1 | 1 - 2 | 617 | 98 | |
GenBank mRNA | AY056111.1 | - | 1 - 2 | 281 | 98 | |
AY072614.1 | - | 1 - 2 | 228 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |