Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338731.1 | NP_001327885.1 | 1 | 1429 | 82 | |
NM_113405.2 | NP_189137.1 | 1 | 1241 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |