Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113514.4 | NP_189238.2 | 9 - 10 | 1452 | 152 | |
GenBank mRNA | AY150429.1 | - | 9 - 10 | 1182 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |