Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113591.4 | NP_566799.1 | 4 - 5 | 950 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |