Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113607.4 | NP_566804.3 | 8 - 9 | 2311 | 62 | |
GenBank mRNA | AF002706.1 | - | 8 - 9 | 2193 | 62 | |
AF026549.1 | - | 8 - 9 | 2173 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |