Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113607.4 | NP_566804.3 | 9 - 10 | 2424 | 89 | |
GenBank mRNA | AF002706.1 | - | 9 - 10 | 2306 | 89 | |
AF026549.1 | - | 9 - 10 | 2286 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |