Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345644.1 | NP_001331903.1 | 1 - 2 | 762 | 84 | |
NM_125853.3 | NP_201262.1 | 1 - 2 | 790 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |