Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126028.2 | NP_001119500.1 | 5 - 6 | 1336 | 86 | |
NM_001345669.1 | NP_001318880.1 | 9 - 10 | 1541 | 86 | ||
NM_125908.5 | NP_201314.3 | 8 - 9 | 1534 | 86 | ||
GenBank mRNA | BT015346.1 | - | 7 - 8 | 1433 | 86 | |
BT015797.1 | - | 7 - 8 | 1371 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |