Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345687.1 | NP_001332735.1 | 4 - 5 | 1342 | 69 | |
NM_125943.2 | NP_201348.1 | 4 - 5 | 1014 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |