Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP77B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101033.4 | NP_172626.1 | 1 | 1602 | 84 | |
GenBank mRNA | AK229331.1 | - | 1 | 1535 | 84 | |
AY084787.1 | - | 1 | 1540 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |