Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084049.2 | NP_001077518.1 | 3 - 4 | 643 | 106 | |
GenBank mRNA | BT002021.1 | - | 3 - 4 | 489 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |