Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332031.1 | NP_001321808.1 | 5 - 6 | 812 | 109 | |
NM_101107.4 | NP_563904.2 | 5 - 6 | 616 | 109 | ||
GenBank mRNA | AK118086.1 | - | 5 - 6 | 540 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |