Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101129.3 | NP_172719.1 | 2 - 3 | 952 | 125 | |
GenBank mRNA | AY057692.1 | - | 2 - 3 | 847 | 125 | |
AY143928.1 | - | 1 - 2 | 598 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |