Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECOY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160866.1 | NP_001154338.1 | 2 - 3 | 334 | 70 | |
NM_101329.4 | NP_172914.1 | 2 - 3 | 438 | 70 | ||
GenBank mRNA | AY127001.1 | - | 2 - 3 | 334 | 70 | |
BT001031.1 | - | 2 - 3 | 231 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |