Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECOY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160866.1 | NP_001154338.1 | 4 - 5 | 542 | 86 | |
NM_101329.4 | NP_172914.1 | 4 - 5 | 646 | 86 | ||
GenBank mRNA | AY127001.1 | - | 4 - 5 | 542 | 86 | |
BT001031.1 | - | 4 - 5 | 439 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |