Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198066.1 | NP_001184995.1 | 19 - 20 | 2107 | 82 | |
NM_101333.5 | NP_172918.2 | 18 - 19 | 2309 | 82 | ||
GenBank mRNA | DQ104324.1 | - | 18 - 19 | 2147 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |