Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035999.3 | NP_001031076.1 | 3 - 4 | 723 | 76 | |
NM_102009.3 | NP_564144.1 | 3 - 4 | 724 | 76 | ||
GenBank mRNA | AF324715.2 | - | 3 - 4 | 679 | 76 | |
AF326894.1 | - | 3 - 4 | 679 | 76 | ||
AF446886.1 | - | 3 - 4 | 659 | 76 | ||
AK317001.1 | - | 3 - 4 | 659 | 76 | ||
AY052666.1 | - | 3 - 4 | 723 | 76 | ||
AY113873.1 | - | 3 - 4 | 659 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |