Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UREF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332530.1 | NP_001322780.1 | 2 | 823 | 68 | |
NM_102032.4 | NP_173602.1 | 1 | 1130 | 68 | ||
GenBank mRNA | AK175196.1 | - | 1 | 1084 | 68 | |
BT012628.1 | - | 1 | 685 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |