Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334652.1 | NP_001323240.1 | 4 - 5 | 397 | 69 | |
NM_106112.3 | NP_177592.1 | 3 - 4 | 513 | 69 | ||
GenBank mRNA | AK118825.1 | - | 3 - 4 | 272 | 69 | |
BT005492.1 | - | 3 - 4 | 218 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |