Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106118.3 | NP_177598.1 | 1 - 2 | 517 | 67 | |
GenBank mRNA | AK117614.1 | - | 1 - 2 | 383 | 67 | |
AY088052.1 | - | 1 - 2 | 383 | 67 | ||
BT005419.1 | - | 1 - 2 | 323 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |