Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106147.2 | NP_177627.1 | 4 - 5 | 660 | 122 | |
GenBank mRNA | AF305720.1 | - | 4 - 5 | 345 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |