Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334788.1 | NP_001321320.1 | 7 - 8 | 1790 | 77 | |
NM_106407.7 | NP_177882.1 | 8 - 9 | 1717 | 77 | ||
GenBank mRNA | AF503759.1 | - | 7 - 8 | 1442 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |