Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334800.1 | NP_001322496.1 | 1 - 2 | 1262 | 85 | |
NM_106434.2 | NP_565161.1 | 1 - 2 | 1393 | 85 | ||
GenBank mRNA | BT002749.1 | - | 1 - 2 | 1062 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |