Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334801.1 | NP_001321009.1 | 1 - 2 | 331 | 101 | |
NM_106435.2 | NP_177909.2 | 1 - 2 | 309 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |