Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161003.2 | NP_001154475.1 | 3 - 4 | 1121 | 141 | |
NM_106474.5 | NP_177948.3 | 2 - 3 | 1236 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |