Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106498.4 | NP_177972.2 | 1 - 2 | 508 | 100 | |
GenBank mRNA | AB071514.2 | - | 1 - 2 | 353 | 100 | |
AB188497.1 | - | 1 - 2 | 189 | 100 | ||
BT025550.1 | - | 1 - 2 | 189 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |