Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106592.5 | NP_178062.1 | 4 - 5 | 499 | 103 | |
GenBank mRNA | AF117335.1 | - | 4 - 5 | 417 | 103 | |
AF428367.1 | - | 4 - 5 | 419 | 103 | ||
AY056147.1 | - | 4 - 5 | 405 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |