Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334951.1 | NP_001320554.1 | 2 - 3 | 598 | 107 | |
NM_106654.3 | NP_178122.1 | 3 - 4 | 1040 | 107 | ||
GenBank mRNA | AY072021.1 | - | 3 - 4 | 354 | 107 | |
BT001122.1 | - | 3 - 4 | 291 | 107 | ||
X96866.1 | - | 313 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |