Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334962.1 | NP_001323118.1 | 2 - 3 | 836 | 99 | |
NM_001334963.1 | NP_001323117.1 | 3 - 4 | 500 | 99 | ||
NM_001334964.1 | NP_001323120.1 | 2 - 3 | 590 | 99 | ||
NM_001334965.1 | NP_001323116.1 | 2 - 3 | 576 | 99 | ||
NM_001334966.1 | NP_001323119.1 | 3 - 4 | 468 | 99 | ||
NM_106668.6 | NP_178136.1 | 3 - 4 | 483 | 99 | ||
GenBank mRNA | AK175552.1 | - | 3 - 4 | 433 | 99 | |
BT010484.1 | - | 2 - 3 | 141 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |