Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335016.1 | NP_001321606.1 | 6 - 7 | 649 | 67 | |
NM_106731.3 | NP_178198.1 | 5 - 6 | 846 | 67 | ||
GenBank mRNA | AF165125.1 | - | 5 - 6 | 492 | 67 | |
BT029300.1 | - | 5 - 6 | 425 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |