Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G09100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111745.3 | NP_187522.2 | 4 - 5 | 722 | 103 | |
NM_202534.2 | NP_974263.1 | 4 - 5 | 737 | 103 | ||
GenBank mRNA | AY125498.1 | - | 4 - 5 | 716 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |