Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111764.4 | NP_187541.1 | 4 - 5 | 617 | 100 | |
GenBank mRNA | AY062703.1 | - | 4 - 5 | 582 | 100 | |
AY093373.1 | - | 4 - 5 | 396 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |