Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDHAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125135.1 | NP_001118607.1 | 7 - 8 | 870 | 69 | |
NM_111829.4 | NP_566361.1 | 7 - 8 | 990 | 69 | ||
GenBank mRNA | AY084556.1 | - | 7 - 8 | 892 | 69 | |
AY093765.1 | - | 7 - 8 | 894 | 69 | ||
BT001054.1 | - | 7 - 8 | 849 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |