Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111872.6 | NP_187648.1 | 2 - 3 | 1005 | 71 | |
GenBank mRNA | AY063863.1 | - | 2 - 3 | 930 | 71 | |
AY096363.1 | - | 2 - 3 | 842 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |