Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111953.3 | NP_187727.1 | 3 - 4 | 1075 | 76 | |
GenBank mRNA | D26019.1 | - | 3 - 4 | 784 | 76 | |
L22961.1 | - | 3 - 4 | 662 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |