Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084749.1 | NP_001078218.1 | 6 - 7 | 932 | 60 | |
NM_113690.3 | NP_566824.1 | 8 - 9 | 1217 | 60 | ||
GenBank mRNA | AY046004.1 | - | 8 - 9 | 1148 | 60 | |
AY079315.1 | - | 8 - 9 | 1074 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |