Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114472.3 | NP_190189.1 | 1 | 614 | 98 | |
GenBank mRNA | AY084352.1 | - | 1 | 614 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |