Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114549.4 | NP_190266.1 | 3 - 4 | 1230 | 152 | |
GenBank mRNA | AK227072.1 | - | 3 - 4 | 1152 | 152 | |
M93412.1 | - | 3 - 4 | 941 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |