Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114564.3 | NP_190281.1 | 4 - 5 | 971 | 108 | |
GenBank mRNA | AY090236.1 | - | 4 - 5 | 692 | 108 | |
BT003012.1 | - | 3 - 4 | 560 | 108 | ||
KJ138797.1 | - | 3 - 4 | 560 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |