Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339291.1 | NP_001319701.1 | 8 - 9 | 1628 | 124 | |
NM_001339292.1 | NP_001326351.1 | 8 - 9 | 1769 | 124 | ||
NM_001339296.1 | NP_001326350.1 | 7 - 8 | 1751 | 124 | ||
NM_114565.3 | NP_190282.2 | 8 - 9 | 1673 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |