Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114604.2 | NP_190320.1 | 5 - 6 | 984 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |