Search
Search

ALDH10A9| Gene Symbol: ALDH10A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114686.4 | NP_190400.1 | 14 - 15 | 1529 | 66 | |
| GenBank mRNA | AF370333.1 | - | 14 - 15 | 1477 | 66 | |
| AY062987.1 | - | 14 - 15 | 1421 | 66 | ||
Set Membership: |
|