Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339453.1 | NP_001327187.1 | 3 - 4 | 396 | 134 | |
NM_114870.4 | NP_190579.2 | 4 - 5 | 471 | 134 | ||
GenBank mRNA | BT030315.1 | - | 4 - 5 | 366 | 134 | |
EU682309.1 | - | 4 - 5 | 366 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |