Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KICP-02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339461.1 | NP_001326509.1 | 4 - 5 | 846 | 78 | |
NM_001339462.1 | NP_001326510.1 | 3 - 4 | 888 | 78 | ||
NM_001339463.1 | NP_001326508.1 | 4 - 5 | 846 | 78 | ||
NM_114884.5 | NP_566931.1 | 4 - 5 | 846 | 78 | ||
GenBank mRNA | AB061676.1 | - | 4 - 5 | 821 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |