Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117144.4 | NP_192814.1 | 1 - 2 | 1144 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |