Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117144.4 | NP_192814.1 | 5 - 6 | 1876 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |