Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340732.1 | NP_001328454.1 | 10 - 11 | 1941 | 75 | |
NM_117243.4 | NP_567380.2 | 10 - 11 | 1874 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |